Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Rabbit anti-Human, Alexa Fluor™ 750, Clone: C225 (Cetuximab), Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252671AF750

Catalog No. NP22671A70


Add to cart

Description

Description

EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry.
Specifications

Specifications

EGFR
C225 (Cetuximab)
Flow Cytometry, ELISA, Immunohistochemistry
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Recognises human EGFR.
ELISA, Flow Cytometry, Immunohistochemistry
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
EGFR
The epidermal growth factor receptor is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands.
0.1 mL
Primary
1956
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit