Learn More
EGFR Rabbit anti-Human,Mouse, KHL, Clone: 11E19, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB0433825UL
 View more versions of this product
Catalog No. ZRB043382MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
EGFR Monoclonal antibody specifically detects EGFR in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), RNAi/siRNA/Gene Knockdown, Western Blotting.
Specifications
Specifications
EGFR
11E19
KHL
Epidermal growth factor receptor;EC: 2.7.10.1;Proto-oncogene c-ErbB-1;Receptor tyrosine-protein kinase erbB-1
Rabbit
IgG
Protein A Purified
Signaling
Primary
Human, Mouse
Immunocytochemistry, Immunohistochemistry (Paraffin), Knockdown, Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Purified
EGFR, ERBB, ERBB1, HER1
KLH-conjugated linear peptide corresponding to 30 amino acids from the C-terminal region of human Epidermal growth factor receptor.
∼170 kDa observed; 134.28 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal