Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Rabbit anti-Human,Mouse, KHL, Clone: 11E19, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen EGFR
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Knockdown, Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB043382MI
View Documents
MilliporeSigma
ZRB0433825UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB043384MI
View Documents
MilliporeSigma
ZRB043384X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

EGFR Monoclonal antibody specifically detects EGFR in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), RNAi/siRNA/Gene Knockdown, Western Blotting.
Specifications

Specifications

EGFR
KHL
Rabbit
Epidermal growth factor receptor;EC: 2.7.10.1;Proto-oncogene c-ErbB-1;Receptor tyrosine-protein kinase erbB-1
KLH-conjugated linear peptide corresponding to 30 amino acids from the C-terminal region of human Epidermal growth factor receptor.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Knockdown, Western Blot
Purified
Human, Mouse
EGFR, ERBB, ERBB1, HER1
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit