Learn More
EGFR Rabbit anti-Human, Unconjugated, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation DH0083
Catalog No. 89-339-483
Add to cart
Description
Description
EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for IF.
Specifications
Specifications
EGFR
0.03 mg/mL
epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian)
Liquid
Rabbit
IgG
Store at -20°C. Do not freeze.
Monoclonal
Human
Immunofluorescence
Unconjugated
EGFR
ERBB, ERBB1, HER1, PIG61, mENA
Human recombinant EGFR
17 μL
Primary
1956