Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Rabbit anti-Human, Unconjugated, Clone: GFR/2968R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00544

Catalog No. 89-339-500


Add to cart

Description

Description

EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for IHC-P.
Specifications

Specifications

EGFR
GFR/2968R
epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian)
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
EGFR
ERBB, ERBB1, HER1, PIG61, mENA
Recombinant protein corresponding to full-length human EGFR
100 μg
Primary
1956
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit