Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Rat anti-Mouse, DyLight 350, Clone: 5C14, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11094153UV

Catalog No. NBT94153UV


Add to cart

Description

Description

EGFR Monoclonal antibody specifically detects EGFR in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

EGFR
5C14
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
EGFR
Recombinant Mouse EGF-R extracellular domain
Protein G purified
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases
Primary
1956
Mouse
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This recognizes mouse EGF-R in western blot.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit