Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Egg Shaped Stir Bar
United Scientific Egg Shaped Stir Bar for use with Hot Plate/Stirrers.
Supplier: United Scientific™ MSHL50
Description
- Egg-shaped magnetic stir bar with blunt edges
- High-quality magnet coated with PTFE
- Available in multiple sizes and designs
Specifications
20 mm | |
White | |
Stir Bar | |
Egg |
PTFE | |
Autoclavable | |
50 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction