Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGLN1/PHD2 Antibody (2445D), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP278782DL594
Description
EGLN1/PHD2 Monoclonal specifically detects EGLN1/PHD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EGLN1/PHD2 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EGLN1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human EGLN1/PHD2 (amino acids 2-426) [UniProt #Q9QZT9] | |
0.1 mL | |
Cancer, Chromatin Research, Hypoxia | |
Human, Mouse |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2445D | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179 | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction