Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGLN1/PHD2, Mouse anti-Human, Clone: 366G/76/3, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP280712
Description
EGLN1/PHD2 Monoclonal antibody specifically detects EGLN1/PHD2 in Human Samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
EGLN1/PHD2 | |
366G/76/3 | |
Unconjugated | |
Tris-Glycine, 0.15 M NaCl | |
C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179 | |
Mouse | |
IgG1 | |
0.1 mL | |
Cancer, Chromatin Research, Hypoxia | |
Primary | |
54583 |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Western Blot 0.5-2.0 ug/ml, Simple Western 0.0763888888888889, Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:400 | |
Purified | |
EGLN1 | |
A peptide within residues 1-50 of human PHD2/HIF Prolyl Hydroxylase 2. [Swiss-Prot# Q9GZT9] | |
Protein G purified | |
RUO | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title