Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN1/PHD2, Rabbit anti-Human,Mouse, Alexa Fluor 405, Clone: 2445D, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP278782AF405

Catalog No. NB009428


Add to cart

Description

Description

EGLN1/PHD2 Monoclonal antibody specifically detects EGLN1/PHD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

EGLN1/PHD2
2445D
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
EGLN1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human EGLN1/PHD2 (amino acids 2-426) [UniProt #Q9QZT9]
0.1 mL
Cancer, Chromatin Research, Hypoxia
Primary
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179
Rabbit
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit