Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN2/PHD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259179

Catalog No. NBP259179

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

EGLN2/PHD1 Polyclonal antibody specifically detects EGLN2/PHD1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

EGLN2/PHD1
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
DKFZp434E026, EC 1.14.11, EC 1.14.11.-, egl nine homolog 2, egl nine homolog 2 (C. elegans), EIT6, Estrogen-induced tag 6, FLJ95603, HIF-PH1, HIFPH1 EGL nine (C.elegans) homolog 2, HIF-prolyl hydroxylase 1, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 1, PHD1, Prolyl hydroxylase domain-containing protein 1
Rabbit
IgG
100 ul
Cancer, Hypoxia, Signal Transduction
Primary
112398
Immunoprecipitation, Western Blot
Western Blot 1:1000, Immunoprecipitation 8 - 15 ul/mg lysate
Affinity Purified
EGLN2
Maps to a region between 1 and 50
Immunogen affinity purified
RUO
Store at 4C.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only