Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN3 293T Cell Overexpression Lysate (Denatured), Abnova

Cell transient overexpression lysate for EGLN3

Manufacturer:  Abnova Corporation H00112399T01

Catalog No. 89-009-667


Add to cart

Description

Description

  • egl nine homolog 3 (C. elegans)
  • Transfected cell line: 293T
  • Plasmid: pCMV-EGLN3 full-length
  • Host: Human
  • Theoretical molecular weight: 26.4kDa
  • Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)

Western Blotting

Specifications

Specifications

293T
NM_022073
6.8
Transient overexpression cell Lysate was tested with Anti-EGLN3 antibody (H00112399-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
EGLN3 293T Cell Transient Overexpression Lysate(Denatured)
26.4
pCMV-EGLN3 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit