Learn More
Abnova™ EGLN3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00112399T03
Catalog No. 89-942-947
Add to cart
Specifications
Specifications
Western Blot
293T
NM_022073
pCMV-EGLN3 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
EGLN3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 27.3kDa
Transient overexpression cell lysate was tested with anti-EGLN3 antibody (Mfr. No. H00112399-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)