Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLU, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

170984-72-2
C7H13NO4
175.184
QFYBYZLHPIALCZ-ZETCQYMHSA-N
(2S)-α-Ethylglutamic acid
5311079
(2S)-2-amino-2-ethylpentanedioic acid
CCC(CCC(=O)O)(C(=O)O)N

Highly selective group II mGlu antagonist

Selective antagonist of presynaptically-mediated (1S,3S)-ACPD-induced depression of motoneuron excitation in neonatal rat spinal cord; presumed group II mGlu receptor antagonist. Also available as part of the Group II mGlu Receptor Tocriset™ .

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Formula Weight 175.18
Quantity 10mg
Chemical Name or Material EGLU
Grade >95%

Recommended Storage : Store at -20°C