Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGR2 Antibody (OTI1B12), HRP, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270624H

Catalog No. NBP270624H


Add to Cart

Description

Description

EGR2 Monoclonal specifically detects EGR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

EGR2
Monoclonal
HRP
PBS with No Preservative
EGR2
Recombinant protein fragment corresponding to amino acids 156-476 of human EGR2 (NP_000390) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunohistochemistry
OTI1B12
Western Blot, Immunohistochemistry
AT591, CMT1D, CMT4E, early growth response 2, early growth response 2 (Krox-20 homolog, Drosophila), early growth response protein 2, EGR-2, FLJ14547, Krox-20 homolog, Drosophila, KROX-20, Drosophila, homolog (early growth response-2), KROX20DKFZp686J1957, Zinc finger protein Krox-20
Mouse
Affinity Purified
Chromatin Research, Neurodegeneration, Neuroscience, Transcription Factors and Regulators, Zinc Finger
1959.0
Store at 4°C in the dark.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit