Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGR4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550496

Catalog No. PA550496


Add to cart

Description

Description

The antibody detects endogenous levels of total EGR4 protein.

Early growth response protein 4 (EGR-4), also known as AT133, is a protein that in humans is encoded by the EGR4 gene. EGR-4 is a member of the early growth response (EGF) family of zinc finger transcription factors. Transcriptional regulator. Recognizes and binds to the DNA sequence 5 ft.-GCGGGGGCG-3 ft. (GSG). Activates the transcription of target genes whose products are required for mitogenesis and differentiation.
Specifications

Specifications

EGR4
Polyclonal
Unconjugated
EGR4
Liquid
EGR4
Synthetic peptide corresponding to a region derived from internal residues of human early growth response 4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.8 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q05215, Q9WUF2
NGFIC, NGFI-C, PAT133
Rabbit
IgG
100 μL
-20°C
Primary
13656, 1961
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit