Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EHD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549403

Catalog No. PA549403


Add to cart

Description

Description

This gene encodes a member of the EH domain-containing protein family. These proteins are characterized by a C-terminal EF-hand domain, a nucleotide-binding consensus site at the N terminus and a bipartite nuclear localization signal. The encoded protein interacts with the actin cytoskeleton through an N-terminal domain and also binds to an EH domain-binding protein through the C-terminal EH domain. This interaction appears to connect clathrin-dependent endocytosis to actin, suggesting that this gene product participates in the endocytic pathway.
Specifications

Specifications

EHD2
Polyclonal
PBS with 0.09% sodium azide
Q9NZN4, Q8BH64, Q4V8H8
EH domain-containing protein 2, PAST homolog 2, EHD2, PAST2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
259300, 30846, 361512
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
EHD2
Liquid
EHD2
KLH conjugated synthetic peptide between 415-449 amino acids from the C-terminal region of human EHD2.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit