Learn More
Abnova™ EHD4 HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Human overexpression lysate
Manufacturer:  Abnova™ L082T6
Catalog No. 89-942-963
Add to cart
Specifications
Specifications
2 mg/ml
Human
EHD4 HEK293 Cell Transient Overexpression Lysate(Non-Denatured)
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF.
pCMV-EHD4 full length ;pCMV-EHD4 full length
100 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Immunoprecipitation, Western Blot
HEK293
BC006287
Theoretical M.W. 61kDa
Transient overexpression cell lysate was tested with anti-EHD4 antibody (Mfr. No. H00030844-M01) by Western Blot.
In modified RIPA Lysis Buffer.