Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EHF Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen EHF
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125968
View Documents
Novus Biologicals
NBP31053325UL
25 μg
Each for $106.19
Add to cart
 
NB125967
View Documents
Novus Biologicals
NBP310533100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

EHF Polyclonal antibody specifically detects EHF in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

EHF
Western Blot, Immunohistochemistry
Purified
RUO
PBS buffer, 2% sucrose
26298
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Immunohistochemistry
Unconjugated
Rabbit
Human, Mouse
Epithelium-specific Ets transcription factor 3, ESE-3, ESE3 transcription factor, ESE3B, ESE3hEHF, ESEJepithelium-specific ets factor 3, ETS domain-containing transcription factor, ets homologous factor
The immunogen is a synthetic peptide directed towards the N terminal region of mouse EHF (NP_031940). Peptide sequence SCIPFQEFDISGEHLCSMSLQEFTRAAGSAGQLLYSNLQHLKWNGQCSSD
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit