Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EHMT1 Mouse anti-Human, Clone:1B7B4, Abnova™

Mouse monoclonal antibody raised against recombinant human EHMT1.

Manufacturer:  Abnova Corporation MAB17864

Catalog No. 89-334-507

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is a histone methyltransferase that is part of the E2F6 complex, which represses transcription. The encoded protein methylates the Lys-9 position of histone H3, which tags it for transcriptional repression. This protein may be involved in the silencing of MYC- and E2F-responsive genes and therefore could play a role in the G0/G1 cell cycle transition. Defects in this gene are a cause of chromosome 9q subtelomeric deletion syndrome (9q-syndrome). Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications

Specifications

EHMT1
1B7B4
euchromatic histone-lysine N-methyltransferase 1
In ascitic fluid (0.03% sodium azide)
Liquid
Mouse
IgG1
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Western Blot
Unconjugated
ELISA (1:10000)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
EHMT1
DEL9q34, DKFZp667M072, EUHMTASE1, Eu-HMTase1, FLJ12879, FP13812, GLP, KIAA1876, KMT1D, bA188C12.1
Recombinant protein corresponding to human EHMT1.
100μg
Antibody
79813
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit