Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EI24 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549527

Catalog No. PA549527

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Acts as a negative growth regulator via p53-mediated apoptosis pathway. Regulates formation of degradative autolysosomes during autophagy.
Specifications

Specifications

EI24
Polyclonal
PBS with 0.09% sodium azide
Q61070
Etoposide-induced protein 24 homolog, p53-induced gene 8 protein, EI24, PIG8
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
13663
Western Blot
Unconjugated
Ei24
Liquid
EI24
KLH conjugated synthetic peptide between 27-61 amino acids from human EI24.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit