Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EI24 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260708
Description
EI24 Polyclonal specifically detects EI24 in Human samples. It is validated for Immunoprecipitation.Specifications
EI24 | |
Polyclonal | |
Immunoprecipitation 8-15 ul/mg lysate | |
EPG4, etoposide induced 2.4 mRNA, p53-induced gene 8 protein, PIG8etoposide-induced protein 2.4 homolog, TP53I8, tumor protein p53 inducible protein 8 | |
Rabbit | |
Affinity Purified | |
Apoptosis | |
9538 | |
Store at 4degreesC. |
Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
EI24 | |
between 50 and 100 | |
100 μL | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title