Learn More
EID3 Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS6378R
Catalog No. 50-198-7225
Add to cart
Description
Description
Act as a repressor of nuclear receptor-dependent transcription possibly by interfering with CREBBP-dependent coactivation. May function as a coinhibitor of other CREBBP/EP300-dependent transcription factors.
Specifications
Specifications
EID3
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
EID3
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
493861
EID3
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q8N140
1700027M21Rik; E1A-like inhibitor of differentiation 3; EID-1-like inhibitor of differentiation 3; EID3; EID-3; EP300 interacting inhibitor of differentiation 3; EP300-interacting inhibitor of differentiation 3; non-SMC element 4 homolog B; Non-structural maintenance of chromosomes element 4 homolog B; NS4EB; NSE4B; NSMCE4B; testis tissue sperm-binding protein Li 96mP
KLH conjugated synthetic peptide derived from human EID3.
Protein A
RUO
Antibody
Polyclonal
Human