Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF1 Mouse anti-Human, Clone: OTI2B9, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502844S

Catalog No. 50-167-3855


Add to cart

Description

Description

Necessary for scanning and involved in initiation site selection. Promotes the assembly of 48S ribosomal complexes at the authentic initiation codon of a conventional capped mRNA.
Specifications

Specifications

EIF1
Monoclonal
1.32 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P41567
A121, EIF-1, EIF1A, ISO1, SUI1
Mouse
IgG2a
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10209
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2B9
Unconjugated
EIF1
Liquid
EIF1
Full length human recombit protein of human EIF1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit