Learn More
EIF1AD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 205281AP
Catalog No. 50-173-2938
Add to cart
Specifications
Specifications
EIF1AD
0.54 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3THJ3, Q5RKI6, Q8N9N8
EIF1AD, Haponin
Rabbit
IgG
150 μL
-20°C
Primary
293673, 69860, 84285
Western Blot
Unconjugated
EIF1AD
Liquid
Eif1ad
EIF1AD Fusion Protein Ag14390
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat