Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF1AY Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI2G2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271417AF647

Catalog No. N271417A647


Add to cart

Description

Description

EIF1AY Monoclonal antibody specifically detects EIF1AY in Human samples. It is validated for Western Blot.
Specifications

Specifications

EIF1AY
OTI2G2
Western Blot
Affinity Purified, Conjugated
EIF1AY
Recombinant protein fragment corresponding to amino acids 2-144 of human EIF1AY (NP_004672) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
eIF-1A Y isoform, eIF-4C, eukaryotic translation initiation factor 1A, Y chromosome, eukaryotic translation initiation factor 1A, Y-chromosomal, eukaryotic translation initiation factor 1A, Y-linked, Eukaryotic translation initiation factor 4C
Mouse
IgG2a
0.1 mL
Primary
9086.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit