Learn More
eIF2 alpha/EIF2S1 Antibody (OTI3H4), HRP, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270626H
Catalog No. NBP270626H
Add to Cart
Description
Description
eIF2 alpha/EIF2S1 Monoclonal specifically detects eIF2 alpha/EIF2S1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
eIF2 alpha/EIF2S1
Monoclonal
HRP
PBS with No Preservative
EIF2S1
Full length recombinant protein of human eIF2 alpha/EIF2S1 (NP_004085) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat, Canine, Primate, Monkey
IgG1
Western Blot, Immunohistochemistry
OTI3H4
Western Blot, Immunohistochemistry
EIF2, EIF-2, EIF2A, EIF-2A, EIF-2alpha, EIF2S1 eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa
Mouse
Affinity Purified
Phospho Specific
1965.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions