Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF2 gamma Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100499
Description
Antibody detects endogenous levels of total EIF2G/EIF2S3.
The initiation of protein synthesis in eukaryotic cells is regulated by interactions between protein initiation factors and RNA molecules. The eukaryotic initiation complex is composed of three subunits, designated eIF2 alpha, eIF2 beta and eIF2 gamma (eukaryotic translation initiation factor 2 alpha, beta and gamma, respectively), all of which work in concert to form a ternary complex with GTP and tRNA in the early stages of protein synthesis. eIF2S3 (eukaryotic translation initiation factor 2, subunit 3), also known as eIF2G, is a 472 amino acid protein that belongs to the gamma subfamily of GTP-binding elongation factor proteins. Existing as a heterotrimer composed of an alpha, beta and gamma chain, eIF2S3 functions to bind ribosomal subunits and catalyze the subsequent formation of preinitiation complexes necessary for protein synthesis.
Specifications
eIF2 gamma | |
Polyclonal | |
Unconjugated | |
EIF2S3 | |
EIF2; EIF2G; eIF-2gA; EIF2gamma; eIF-2-gamma; eIF-2-gamma X; eIF-2gX; Eif2s3; Eif2s3x; Eukaryotic translation initiation factor 2 subunit 3; Eukaryotic translation initiation factor 2 subunit 3, X-linked; Eukaryotic translation initiation factor 2 subunit gamma; eukaryotic translation initiation factor 2 subunit gamma X; Eukaryotic translation initiation factor 2 subunit gamma, X-linked; eukaryotic translation initiation factor 2, subunit 3 gamma, 52kDa; eukaryotic translation initiation factor 2, subunit 3, structural gene X-linked; eukaryotic translation initiation factor 2G; PP42 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1968 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P41091 | |
EIF2S3 | |
A synthesized peptide derived from human EIF2S3(Accession P41091), corresponding to amino acid residues A86-P136. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction