Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF2A Mouse anti-Human, PerCP, Clone: 3A7A8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237279PCP

Catalog No. NBP237279W

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

eIF2A
3A7A8
Flow Cytometry
Purified
EIF2A
Purified recombinant fragment of human EIF2A (AA: 448-576) expressed in E. Coli.
Protein A or G purified
Chromatin Research, DNA Repair, Lipid and Metabolism, Phospho Specific, Translation Control, Unfolded Protein Response
Primary
83939
Flow Cytometry
PerCP
PBS with 0.05% Sodium Azide
EIF-2, EIF-2A, EIF2AEIF2, EIF-2alpha, eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa, subunit 1 (alpha, 35kD )
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit