Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EIF2AK1 (HRI), GST Tag Recombinant Protein
Human EIF2AK1 (HRI), GST Tag Recombinant Protein
$558.00
Specifications
Accession Number | NP_055228.2 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 98.6 kDa |
Gene Alias | EIF2AK1 (HRI) |
KinaseFamily | PEK Kinase Family |
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
PV5856
|
LIFE TECHNOLOGIES
PV5856 |
10μg | 10μg |
Each of 1 for $558.00
|
|
||||
Description
EIF2AK1 (HRI) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK1, signaling is initiated by low levels of heme.Specifications
NP_055228.2 | |
98.6 kDa | |
PEK Kinase Family | |
GST Tag | |
Ser⁄Thr Kinases: Other | |
Baculovirus |
Kinase Assay | |
EIF2AK1 (HRI) | |
Human | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Dry Ice | |
Kinases & Inhibitors |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title