Learn More
EIF2AK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 204991AP
Catalog No. 50-173-2922
Add to cart
Description
Description
The HRI gene is localized to 7p22 where its 3 ft. end slightly overlaps the 3 ft. end of the gene JTV1. The two genes are transcribed from opposite strands. Studies in rat and rabbit suggest that the HRI gene product phosphorylates the alpha subunit of eukaryotic initiation factor 2. Its kinase activity is induced by low levels of heme and inhibited by the presence of heme.
Specifications
Specifications
EIF2AK1
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q63185, Q9BQI3, Q9Z2R9
EIF2AK1, HCR, Heme controlled repressor, Heme regulated inhibitor, HRI, KIAA1369
Rabbit
IgG
150 μL
-20°C
Primary
15467, 27102, 27137
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
EIF2AK1
Liquid
Eif2ak1
EIF2AK1 Fusion Protein Ag14236
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat