Learn More
Abnova™ EIF2B2 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00008892T02
Catalog No. 89-943-006
Add to cart
Specifications
Specifications
Western Blot
293T
NM_014239.2
pCMV-EIF2B2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
EIF2B2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 39kDa
Transient overexpression cell lysate was tested with anti-EIF2B2 antibody (Mfr. No. H00008892-D01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)