Sino Biological™ EIF2C1 Recombinant Human Protein, N-His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding full length of human EIF2C1 (Met 1-Ala 857) fused to polyhistidine tag at N-terminus

Manufacturer: Sino Biological™ 11225H07B50

 View more versions of this product

Catalog No. 11-225-H07B50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Purity ≥90% by SDS-PAGE
    Source Human
    Structural Form Full Length, Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name Eukaryotic Translation Initiation Factor 2C 1
    Gene Symbol EIF2C1
    Protein Tag N-His Tag
    Protein Family Other proteins
    Expression System Insect
    Format Lyophilized
    Molecular Weight 99.5kd
    Quantity 50μg
    Validated Application Assay Standard
    Product Type EIF2C1 Recombinant Human Protein
    For Use With (Application) Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
    Gene ID (Entrez) 26523

    Expressed from DNA sequence encoding full length of human EIF2C1 (Met 1-Ala 857) fused to polyhistidine tag at N-terminus

    • Formulation: 50mM Tris, 100mM NaCl, 10% glycerol, 0.5 PMSF, 0.5mM EDTA, pH 8.0

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit