Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF2S1 Mouse anti-Human, Clone: OTI1H1, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA501402
Description
EIF2a (Eukaryotic translation initiation factor 2A) is a heterotrimer composed of three subunits (alpha, beta, and gamma). This translation initiation factor drives binding of initiator methionyl-tRNA to the 40S ribosome in an AUG-dependent manner to form the 43S pre-initiation complex. The polypeptide can be phosphorylated by related protein kinases activated in response to stress. Phosphorylated EIF2A inhibits EIF2B activity and prevents guanine nucleotide exchange.Specifications
EIF2S1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P05198 | |
EIF-2, EIF-2A, EIF-2alpha, EIF2, EIF2A | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
1965 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1H1 | |
Unconjugated | |
EIF2S1 | |
Liquid | |
EIF2S1 | |
Full length human recombit protein of human EIF2S1 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title