Learn More
EIF3C Mouse anti-Human, Polyclonal, Abnova™
Mouse Polyclonal Antibody.
Manufacturer:  Abnova Corporation H00008663B01P
Catalog No. 89-339-711
Add to cart
Description
Description
EIF3C Polyclonal antibody specifically detects EIF3C in Human, Mouse samples. It is validated for WB-Ce, WB-Tr.
Specifications
Specifications
EIF3C
Unconjugated
EIF3C
EIF3S8, FLJ78287, MGC189744, eIF3-p110
EIF3S8 (NP_003743.1, 1 a.a. ∼ 913 a.a) full-length human protein
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Mouse
Western Blot
eukaryotic translation initiation factor 3, subunit C
NM_003752.3
Mouse
50 μg
Primary
8663