Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF3d Monoclonal Antibody (OTI14D6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526950
Description
eIF3d Monoclonal Antibody for Western Blot, IHC (P)
eIF3D is a protein belonging to the largest of the eukaryotic translation initiation factor family, eukaryotic translation initiation factor-3 (eIF3). eIF3 is a multiprotein complex composed of at least 13 different subunits. eIF3D binds to the 40S ribosomal subunit and thus helps in maintaining the 40S and 60S ribosomal subunits in a dissociated state. An important role of eIF3D is the formation of the 40S initiation complex by interacting with the ternary complex of eIF2/GTP/methionyl-tRNA, and promotes mRNA binding. eIF3D is the major RNA binding subunit of the eIF3 complex. It associates with the subunit p170 of eIF-3. Ubiquitous expression of eIF3D is seen in most of the tissues and it is phosphorylated upon DNA damage, likely by ATM or ATR.
Specifications
eIF3d | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15371 | |
EIF3D | |
Full length human recombinant protein of EIF3D produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI14D6 | |
Unconjugated | |
EIF3D | |
66/67kDa; AA407891; actinin alpha 4; ACTININ-4; Actn4; alpha actinin 4; alpha-actinin-4; eIF3 p66; EIF3D; eIF3d {ECO:0000255; eIF3p66; eIF3-p66; Eif3s7; eIF-3-zeta; eIF3-zeta; eIF-3-zeta {ECO:0000255; eukaryotic translation initiation factor 3 subunit 7; eukaryotic translation initiation factor 3 subunit 7 {ECO:0000255; Eukaryotic translation initiation factor 3 subunit D; eukaryotic translation initiation factor 3 subunit D {ECO:0000255; eukaryotic translation initiation factor 3, subunit 7 (zeta); eukaryotic translation initiation factor 3, subunit 7 zeta; eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa; eukaryotic translation initiation factor 3, subunit D; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; FSGS; FSGS1; HAMAP-Rule:MF_03003}; non-muscle alpha-actinin 4; RP5-1119A7.12-003; si:dkey-165I8.6; translation initiation factor eIF3 p66; translation initiation factor eIF3 p66 subunit; wu:fb12a03; zgc:64097 | |
Mouse | |
Affinity chromatography | |
RUO | |
8664 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction