Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF3d Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA564108

Catalog No. PIPA564108


Add to cart

Description

Description

Immunogen sequence: IVRCEHDGVM TGANGEVSFI NIKTLNEWDS RHCNGVDWRQ KLDSQRGAVI ATELKNNSYK LARWTCCALL AGSEYLKLGY VSRYHVKDSS RHVILGTQQF KPNEFASQIN LSVENAWGIL RCVIDICMKL EEGKYLILKD PNKQVIRVYS LPDGTFSSDE DEEE Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

eIF3D is a protein belonging to the largest of the eukaryotic translation initiation factor family, eukaryotic translation initiation factor-3 (eIF3). eIF3 is a multiprotein complex composed of at least 13 different subunits. eIF3D binds to the 40S ribosomal subunit and thus helps in maintaining the 40S and 60S ribosomal subunits in a dissociated state. An important role of eIF3D is the formation of the 40S initiation complex by interacting with the ternary complex of eIF2/GTP/methionyl-tRNA, and promotes mRNA binding. eIF3D is the major RNA binding subunit of the eIF3 complex. It associates with the subunit p170 of eIF-3. Ubiquitous expression of eIF3D is seen in most of the tissues and it is phosphorylated upon DNA damage, likely by ATM or ATR.
Specifications

Specifications

eIF3d
Polyclonal
Unconjugated
EIF3D
Liquid
EIF3D
Recombit protein corresponding to Human eIF3d
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O15371
RP5-1119A7.12-003, EIF3S7, eIF3-p66, eIF3-zeta, eIF-3-zeta, eIF3 p66, eukaryotic translation initiation factor 3 subunit 7, eukaryotic translation initiation factor 3 subunit D, eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa, translation initiation factor eIF3 p66 subunit
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
8664
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only