Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF3G Rabbit anti-Human, Mouse, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

Manufacturer:  Abnova Corporation PAB29909

Catalog No. 89-033-197


Add to cart

Description

Description

Rabbit polyclonal antibody raised against partial synthetic protein of human EIF3G.

  • High specificity
  • High reactivity
Specifications

Specifications

EIF3G
Unconjugated
Immunohistochemistry (1:250) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user.
O75821
EIF3-P42|EIF3S4|eIF3-delta|eIF3-p44
Rabbit
IgG
100 uL
Store at 4°C for up to one week. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Mouse
Immunohistochemistry, Immunoprecipitation, Western Blot
Rabbit polyclonal antibody raised against partial synthetic protein of human EIF3G.
In 1X PBS , pH 7.4 (2% sucrose, 0.09% sodium azide).
Liquid
EIF3G
A synthetic peptide corresponding to amino acids 218-267 of human EIF3G.
Affinity purification
RUO
Primary
8666
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)