Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4A2 Mouse anti-Human, Clone: PCRP-EIF4A2-2B5, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen eIF4A2
Clone PCRP-EIF4A2-2B5
Concentration 0.2 mg/ml
Dilution ELISA, Protein Array
Applications ELISA, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121753
View Documents
Novus Biologicals
NBP30714010UG
100 μg
Each for $352.89
Add to cart
 
NB121754
View Documents
Novus Biologicals
NBP30714020UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

eIF4A2 Monoclonal antibody specifically detects eIF4A2 in Human samples. It is validated for ELISA, Protein Array
Specifications

Specifications

eIF4A2
0.2 mg/ml
ELISA, Peptide Array
Purified
RUO
Human
ATP-dependent RNA helicase eIF4A-2, BM-010, DDX2Beukaryotic translation initiation factor 4A, isoform 2, EC 3.6.1, EC 3.6.4.13, EIF4A, eIF-4A-II, eIF4A-II, EIF4F, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A2
Recombinant full-length human eIF4A2 protein
Monoclonal
Protein A or G purified
PCRP-EIF4A2-2B5
ELISA, Protein Array
Unconjugated
Mouse
Cancer, mTOR Pathway
10 mM PBS with 0.05% BSA
1974
IgG2a
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit