Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF4B Monoclonal Antibody (355CT12.6.5)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537534
Description
eIF4B Monoclonal Antibody for Western Blot
The eukaryotic translation initiation factor 4B (eIF4B) plays a critical role in recruiting the 40S ribosomal subunit to the mRNA. It functions in close association with eIF4F and eIF4A. It binds near the 5'-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F.
Specifications
eIF4B | |
Monoclonal | |
0.25 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P23588 | |
Eif4b | |
EIF4B recombinant protein. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Western Blot | |
355CT12.6.5 | |
Unconjugated | |
Eif4b | |
2310046H11Rik; AL024095; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; hypothetical protein; PRO1843 | |
Mouse | |
Protein G | |
RUO | |
1975 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction