Learn More
Invitrogen™ EIF4E1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114259
Description
Immunogen sequence: CDYALFKDGI QPMWEDSRNK RGGRWLVSLA KQQRHIELDR LWLETLLCLI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 93%; Zebrafish: 85%.
Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structure.
Specifications
EIF4E1B | |
Polyclonal | |
Unconjugated | |
EIF4E1B | |
EIF4E1B; eukaryotic translation initiation factor 4E family member 1B; eukaryotic translation initiation factor 4E type 1B | |
Rabbit | |
Affinity chromatography | |
RUO | |
253314 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A6NMX2 | |
EIF4E1B | |
Synthetic peptide directed towards the middle region of Human EIF4E1B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.