Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF4E2 Mouse anti-Human, Clone: OTI1F11, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502403

Catalog No. 50-167-3463


Add to cart

Description

Description

Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.
Specifications

Specifications

EIF4E2
Monoclonal
0.54 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O60573
4E-LP, 4EHP, EIF4EL3, IF4e, h4EHP
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9470
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1F11
Unconjugated
EIF4E2
Liquid
EIF4E2
Full length human recombit protein of human EIF4E2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit