Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF4E2 Mouse anti-Human, PE/Atto594, Clone: OTI1F11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271468PEATT594

Catalog No. N271468P594


Add to cart

Description

Description

EIF4E2 Monoclonal antibody specifically detects EIF4E2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

EIF4E2
OTI1F11
Flow Cytometry
Affinity Purified, Conjugated
EIF4E2
Full length recombinant protein of human EIF4E2 (NP_004837) produced in HEK293T cells.
Immunogen affinity purified
DNA Repair, DNA replication Transcription Translation and Splicing
Primary
9470.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
4EHP, 4E-LP, eIF4E type 2, eIF-4E type 2, EIF4EL3, eIF4E-like cap-binding protein, eIF4E-like protein 4E-LP, eukaryotic translation initiation factor 4E family member 2, Eukaryotic translation initiation factor 4E homologous protein, eukaryotic translation initiation factor 4E member 2, eukaryotic translation initiation factor 4E type 2, Eukaryotic translation initiation factor 4E-like 34E-LP, IF4e, mRNA cap-binding protein 4EHP, mRNA cap-binding protein type 3
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit