Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4E2 Mouse anti-Human, Clone: OTI1F2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525413

Catalog No. PIMA525413


Add to cart

Description

Description

EIF4E2 is expressed exclusively in the cytoplasm. This protein recognizes and binds the 7 methylguanosine containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.
Specifications

Specifications

eIF4E2
Monoclonal
0.66 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O60573
Eukaryotic translation initiation factor 4E type 2; eIF4E type 2; eIF-4E type 2; mRNA cap-binding protein type 3; Eukaryotic translation initiation factor 4E-like 3; Eukaryotic translation initiation factor 4E homologous protein; mRNA cap-binding protein 4EHP; eIF4E-like protein 4E-LP; EIF4E2; EIF4EL3;
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9470
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1F2
Unconjugated
EIF4E2
Liquid
EIF4E2
Full length human recombit protein of EIF4E2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit