Learn More
Aviva Systems Biology Corp EIF5 Antibody - N-terminal region (ARP53443_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53443_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EIF5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EIF5. Alternate names: EIF-5AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61026