Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF5 Antibody (6D6H11) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP261762

Catalog No. NBP2-61762


Add to Cart

Description

Description

EIF5 Monoclonal specifically detects EIF5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

EIF5
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Paraffin 1:200-1:1000
eIF-5, EIF-5A, eukaryotic translation initiation factor 5
Mouse
49.2 kDa
0.1 mL
1983
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
6D6H11
Unconjugated
PBS with 0.05% Sodium Azide
EIF5
Purified recombinant fragment of human EIF5 (AA: 1-300) expressed in E. Coli.
Protein G purified
Primary
Human
Purified
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit