Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Automatic Self-Zeroing Buret
SCI_ED

Self-supporting, closed system for classroom and lab use.

Supplier:  Eisco™ BTAUT50

Catalog No. S12804


Add to cart

Description

Description

  • Capacity 50mL (buret) and 500mL (reservoir)
  • Polyethylene base
  • Resolution of 0.1mL
  • With permanent blue markings against white background
  • Pushbutton style stopcock
  • With reservoir, tube and base

Specifications

Specifications

Borosilicate Glass
Automatic stopcock with screw setting and release, 500 mL reservoir, Self-supporting base, Polyethylene filling tube to connect buret to reservoir, Flexible buret tip, Reservoir screw cap with fitting for polyethylene tube
500 mL
50 mL
Flexible Tip
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit