Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Bunsen Burners, Micro

Allows air ratio adjustments

Manufacturer:  Eisco™ CH0994LP/RSD

 View more versions of this product

Catalog No. 17-012-822


Add to cart

Description

Description

  • Smaller version of premium Bunsen burner
  • Expanded base prevents easy tipping, even with hoses connected
  • Spaced feet so burner fits comfortably under stands
  • Carry hot burner with ease and safety using finger handle
  • 8.8cm (3.5 in.) tall
Specifications

Specifications

Micro
4.25 in. (Base)
LPG
88.9 mm
Flame Stabilizer, Handle
1550°C
8 mm
Iron
Safe Continuous Stream of Flammable Gas
3.5 in.
2822°F
4.25 (Base) in.
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit