Learn More
Eisco™ Class B Glass Volumetric Flasks with Polypropylene Stopper Products for Science Education
Eisco™ Class B Glass Volumetric Flasks with Polypropylene Stopper feature one graduation mark, a fitted chemical resistant stopper, and blue printed markings.
Manufacturer:  Eisco™ CH0448C
 View more versions of this product
Catalog No. S13967
Add to cart
Description
Description
  • Made from high quality borosilicate 3.3 glass
  • Include chemical resistant polypropylene stopper
  • Flat bottom design
  • Single graduation mark
  • Blue printed markings
  • Class B, in compliance with ASTM-E288
  • Autoclavable
Specifications
Specifications
No
Autoclavable
Stopper
Borosilicate Glass 3.3
25 mL
Prepare chemical solution
Non-sterile
Flat
White
Clear
Borosilicate Glass
Long