Learn More
Lab-Aids™ Energy Transfer: Waves, Sound and Light Kit
SCI_ED
Produce both longitudinal and transverse mechanical waves and compare their similarities and differences.
$20.25 - $141.00
Specifications
For Use With (Application) Studying wavelengths and frequencies
View More Specs
Includes:
  • Kit contains 28 student worksheets and guides, teacher's guide, six mini wave tanks, six springs, six pieces of elastic string with barbed ends, six plastic paddles, six 15cm rulers, Transparency 1: Wave Characteristics, Transparency 2: Comparing Wavelength
  • Extender materials include mini wave tank, one spring, one piece of elastic string with barbed ends, one plastic paddle, one 15cm ruler
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S98797
SCI_ED
View Documents Promotion Details
Lab-Aids™
211
Waves, Sound and Light Kit
Each for $141.00
Add to cart
S98797A
SCI_ED
View Documents Promotion Details
Lab-Aids™
211EL
Extender Components Kit
Each for $20.25
Add to cart
Description
Description
As an introduction, students use water and springs to observe wavelength and frequency and discover the relationship between them. Their exploration of waves continues through investigating vibrations and the production of sound waves. Using an elasticized string allows students to create and observe different pitch notes and relate the sounds produced to the frequency of the vibrations. They then read about the wave nature of different energy forms, including light and sound, and are introduced to the fundamental features common to all waves. The reading compares and contrasts the similarities and differences among different energy waves and explains how our eyes and ears interpret various wave forms. Accommodates unlimited classes, each with 6 groups of 4 students. Includes Teacher guide, student sheets and assessment sheets.
Order Info
Grades 6 to 12. Enough material for 28 students; extender materials allow more students to participate at the same time.
Specifications
Specifications
Studying wavelengths and frequencies